Search
Search

GSDMC| Gene Symbol: GSDMC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031415.2 | NP_113603.1 | 12 | 2003 | ||
| XM_011517158.1 | XP_011515460.1 | 11 | 1716 | |||
| XM_011517159.1 | XP_011515461.1 | 10 | 1686 | |||
| XM_011517161.1 | XP_011515463.1 | 12 | 1743 | |||
| XM_011517162.1 | XP_011515464.1 | 12 | 1743 | |||
| XM_011517163.2 | XP_011515465.1 | 11 | 970 | |||
| XM_011517165.1 | XP_011515467.1 | 7 | 482 | |||
| XM_011517166.2 | XP_011515468.1 | 8 | 2162 | |||
| XM_017013644.1 | XP_016869133.1 | 12 | 1743 | |||
| XM_017013645.1 | XP_016869134.1 | 10 | 776 | |||
| GenBank mRNA | AB042405.1 | - | 12 | 2012 | ||
| AJ245876.1 | - | 6 | 466 | |||
| BC035321.1 | - | 12 | 1548 | 4753282 | ||
| BC063595.1 | - | 12 | 1531 | 4752106 | ||