Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@39a2e3a0
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Contact Us
Sign In
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
Attention: Existing favorites lists will be migrated to Shared Lists by the end of 2020.
[Learn more about Shared Lists]
LOADING
See other PARD3 SIRNA Assays ›
Gene Symbol:
PARD3
Gene Name:
par-3 family cell polarity regulator
Gene Aliases:
ASIP, Baz, PAR3, PAR3alpha, PARD-3, PARD3A, PPP1R118, SE2-5L16, SE2-5LT1, SE2-5T2
Chromosome Location:
Chr.10: 34109560 - 34815325 on Build GRCh38
UniGene ID:
Hs.131489
Species:
Human
siRNA ID
HSS183488
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PARD3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001184785.1
NP_001171714.1
9
1710
NM_001184786.1
NP_001171715.1
9
1710
NM_001184787.1
NP_001171716.1
9
1710
NM_001184788.1
NP_001171717.1
9
1710
NM_001184789.1
NP_001171718.1
9
1710
NM_001184790.1
NP_001171719.1
8
1578
NM_001184791.1
NP_001171720.1
8
1578
NM_001184792.1
NP_001171721.1
9
1710
NM_001184793.1
NP_001171722.1
9
1710
NM_001184794.1
NP_001171723.1
9
1710
NM_019619.3
NP_062565.2
9
1710
XM_005252528.4
XP_005252585.1
9
1701
XM_005252530.4
XP_005252587.1
9
1701
XM_005252531.4
XP_005252588.1
9
1701
XM_005252532.4
XP_005252589.1
8
1355
XM_005252534.4
XP_005252591.1
9
1701
XM_005252535.4
XP_005252592.1
9
1701
XM_005252536.4
XP_005252593.1
9
1701
XM_011519569.2
XP_011517871.1
9
1701
XM_011519570.2
XP_011517872.1
9
1701
XM_011519571.2
XP_011517873.1
9
1701
XM_011519572.2
XP_011517874.1
9
1701
XM_011519573.2
XP_011517875.1
9
1701
XM_011519575.2
XP_011517877.1
8
1385
XM_011519576.2
XP_011517878.1
8
1356
XM_011519577.2
XP_011517879.1
9
1701
XM_011519578.2
XP_011517880.1
8
1573
XM_011519579.2
XP_011517881.1
7
1254
XM_011519581.2
XP_011517883.1
9
1701
XM_011519583.2
XP_011517885.1
9
1701
XM_011519584.2
XP_011517886.1
9
1701
XM_017016430.1
XP_016871919.1
9
1701
GenBank mRNA
AB073671.1
-
8
1248
AF177228.1
-
2
446
AF196185.1
-
9
1667
AF196186.1
-
9
1667
AF252293.1
-
8
1248
AF332592.1
-
9
1380
AF332593.1
-
9
1380
AF454057.1
-
7
1087
AF454058.1
-
8
1592
AF454059.1
-
5
713
AF467002.1
-
9
1380
AF467003.1
-
9
1380
AF467004.1
-
9
1380
AF467005.1
-
8
1248
AF467006.1
-
9
1380
AK025892.1
-
1
196
AK027735.1
-
7
1007
BC071566.1
-
9
1657
30344732
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-2-prod-green:8080/10.253.226.220:8080.
git-commit: 0f6c802f6da5e59cd1b986a4dc444b4bcf3ac46d
git-url: http://victoria.invitrogen.com:8333/magellan/core.git
git-branch: origin/release/1.15.0-2021.01.02-1.0