Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019644.3 | NP_062618.2 | 3 | 536 | ||
GenBank mRNA | AK302407.1 | - | 3 | 490 | ||
BC007020.2 | - | 1 | 70 | |||
BC032799.1 | - | 3 | 297 | 5170634 | ||
BP369869.1 | - | 3 | 536 | |||
D78334.1 | - | 3 | 292 |