Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CELF4 SIRNA Assays ›
Gene Symbol:
CELF4
Gene Name:
CUGBP, Elav-like family member 4
Gene Aliases:
BRUNOL4, CELF-4
Chromosome Location:
Chr.18: 37243040 - 37566052 on Build GRCh38
UniGene ID:
Hs.435976
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS183572
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CELF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025087.1
NP_001020258.1
4
923
NM_001025088.1
NP_001020259.1
4
920
NM_001025089.1
NP_001020260.1
4
893
NM_020180.3
NP_064565.1
4
923
XM_005258308.3
XP_005258365.1
4
932
XM_011526079.2
XP_011524381.1
4
937
XM_011526081.2
XP_011524383.1
4
771
XM_011526083.2
XP_011524385.1
4
937
XM_011526084.2
XP_011524386.1
4
934
XM_011526085.2
XP_011524387.1
4
938
XM_011526086.2
XP_011524388.1
4
903
XM_011526087.2
XP_011524389.1
4
908
XM_011526090.2
XP_011524392.1
4
935
XM_011526091.2
XP_011524393.1
4
935
XM_011526092.2
XP_011524394.1
4
932
XM_017025840.1
XP_016881329.1
4
770
XM_017025841.1
XP_016881330.1
4
931
XM_017025842.1
XP_016881331.1
4
936
XM_017025843.1
XP_016881332.1
4
933
XM_017025844.1
XP_016881333.1
4
936
XM_017025845.1
XP_016881334.1
4
933
XM_017025846.1
XP_016881335.1
4
904
XM_017025847.1
XP_016881336.1
4
901
XM_017025848.1
XP_016881337.1
4
902
XM_017025849.1
XP_016881338.1
4
902
XM_017025850.1
XP_016881339.1
4
770
XM_017025851.1
XP_016881340.1
4
767
XM_017025852.1
XP_016881341.1
4
770
XM_017025853.1
XP_016881342.1
4
936
XM_017025854.1
XP_016881343.1
4
936
XM_017025855.1
XP_016881344.1
4
933
XM_017025856.1
XP_016881345.1
4
936
XM_017025857.1
XP_016881346.1
4
904
XM_017025858.1
XP_016881347.1
4
771
XM_017025859.1
XP_016881348.1
4
771
XM_017025860.1
XP_016881349.1
4
902
XM_017025861.1
XP_016881350.1
4
770
XM_017025862.1
XP_016881351.1
4
770
XM_017025863.1
XP_016881352.1
4
749
XM_017025864.1
XP_016881353.1
4
741
XM_017025865.1
XP_016881354.1
4
770
XM_017025866.1
XP_016881355.1
4
355
XM_017025867.1
XP_016881356.1
4
332
XM_017025868.1
XP_016881357.1
4
305
XM_017025869.1
XP_016881358.1
4
296
XM_017025870.1
XP_016881359.1
4
305
XM_017025871.1
XP_016881360.1
4
305
XM_017025872.1
XP_016881361.1
4
296
GenBank mRNA
AB209774.1
-
4
717
AF141345.1
-
176
AF248650.1
-
4
632
AF329265.1
-
4
527
AK074596.1
-
4
652
AK126716.1
-
3
177
AK295009.1
-
4
504
BC001946.1
-
4
681
2820541
BC004167.2
-
4
681
2820541
BC045711.1
-
4
580
5261155
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
alternative mRNA splicing, via spliceosome
regulation of alternative mRNA splicing, via spliceosome
mRNA splice site selection
germ cell development
embryo development
negative regulation of translation
negative regulation of mRNA splicing, via spliceosome
positive regulation of mRNA splicing, via spliceosome
negative regulation of excitatory postsynaptic potential
nucleotide binding
translation repressor activity, nucleic acid binding
mRNA binding
BRE binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0