Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284395.1 | NP_001271324.1 | 3 | 469 | ||
NM_001284396.1 | NP_001271325.1 | 3 | 469 | |||
NM_020212.1 | NP_064597.1 | 3 | 469 | |||
XM_006720614.1 | XP_006720677.1 | 3 | 455 | |||
XM_006720615.1 | XP_006720678.1 | 3 | 455 | |||
XM_006720618.2 | XP_006720681.1 | 3 | 455 | |||
XM_011521794.2 | XP_011520096.1 | 3 | 748 | |||
XM_011521795.1 | XP_011520097.1 | 3 | 479 | |||
XM_011521796.2 | XP_011520098.1 | 3 | 455 | |||
XM_011521802.2 | XP_011520104.1 | 3 | 455 | |||
XM_017022415.1 | XP_016877904.1 | 3 | 437 | |||
XM_017022417.1 | XP_016877906.1 | 3 | 455 | |||
GenBank mRNA | AK097525.1 | - | 3 | 437 | ||
AK304683.1 | - | 3 | 455 | |||
BC051026.1 | - | 2 | 422 | |||
BC064626.1 | - | 3 | 469 | 5742386 |