Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLLU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025233.2 | NP_001020404.1 | 1 | 741 | ||
GenBank mRNA | AJ845165.1 | - | 1 | 741 | ||
AJ845166.1 | - | 1 | 741 |