Search
Search

HECW2| Gene Symbol: HECW2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020760.2 | NP_065811.1 | 2 | 418 | ||
| XM_006712646.3 | XP_006712709.1 | 2 | 455 | |||
| XM_017004563.1 | XP_016860052.1 | 2 | 455 | |||
| GenBank mRNA | AB037722.1 | - | 2 | 418 | ||
| BC117194.1 | - | 2 | 334 | 40125745 | ||
| BC117198.1 | - | 2 | 334 | 40125749 | ||