Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020861.2 | NP_065912.1 | 3 | 1397 | ||
XM_005267076.2 | XP_005267133.1 | 3 | 1258 | |||
XM_011536004.2 | XP_011534306.1 | 3 | 1479 | |||
XM_017011137.1 | XP_016866626.1 | 4 | 1657 | |||
GenBank mRNA | AB040916.1 | - | 1 | 848 | ||
AJ420526.1 | - | 1 | 633 | |||
AK092886.1 | - | 4 | 1163 | |||
AK291942.1 | - | 3 | 1340 | |||
AL832957.1 | - | 1 | 1169 | |||
BC020172.1 | - | 3 | 1239 | 4577244 |