Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BACH2 SIRNA Assays ›
Gene Symbol:
BACH2
Gene Name:
BTB domain and CNC homolog 2
Gene Aliases:
BTBD25
Chromosome Location:
Chr.6: 89926528 - 90296912 on Build GRCh38
UniGene ID:
Hs.269764
Species:
Human
siRNA ID
HSS184314
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BACH2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170794.1
NP_001164265.1
5
1263
NM_021813.3
NP_068585.1
7
1434
XM_005248758.4
XP_005248815.1
4
2216
XM_005248759.4
XP_005248816.1
3
2136
XM_011536039.2
XP_011534341.1
6
1345
XM_011536040.2
XP_011534342.1
5
1294
XM_017011156.1
XP_016866645.1
10
2406
XM_017011157.1
XP_016866646.1
9
2212
XM_017011158.1
XP_016866647.1
11
2485
XM_017011159.1
XP_016866648.1
12
2574
XM_017011160.1
XP_016866649.1
9
2328
XM_017011161.1
XP_016866650.1
11
2036
XM_017011162.1
XP_016866651.1
8
2235
XM_017011163.1
XP_016866652.1
10
2407
XM_017011164.1
XP_016866653.1
9
2313
XM_017011165.1
XP_016866654.1
6
1360
XM_017011166.1
XP_016866655.1
8
1518
XM_017011167.1
XP_016866656.1
10
2406
GenBank mRNA
AB208889.1
-
5
900
AF357835.1
-
4
759
AJ271878.1
-
7
1268
AK309526.1
-
7
1565
HE578167.1
-
1
307
HE578168.1
-
1
309
HE578169.1
-
1
313
HE578170.1
-
1
309
HE578171.1
-
1
310
HE578172.1
-
1
309
HE578173.1
-
1
310
HE578174.1
-
1
309
HE578175.1
-
1
312
HE578176.1
-
1
318
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
protein ubiquitination
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription factor activity, sequence-specific DNA binding
ubiquitin-protein transferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-bsnw9:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0