This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
EEFSEC
- Gene Name:
-
eukaryotic elongation factor, selenocysteine-tRNA specific
- Gene Aliases:
-
EFSEC, SELB
- Chromosome Location:
-
Chr.3: 128153456 - 128426193 on Build GRCh38
- UniGene ID:
-
Hs.477498
- Species:
- Human
- Species Specific ID (Flybase ID):
-
-
siRNA Details
|
Gene Symbol:
EEFSEC |
| Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
| RefSeq |
NM_021937.4 |
NP_068756.2 |
1, 2
|
378
|
|
| XM_005247696.2 |
XP_005247753.1 |
1, 2
|
379
|
|
| XM_017006975.1 |
XP_016862464.1 |
1, 2
|
335
|
|
| XM_017006976.1 |
XP_016862465.1 |
1, 2
|
335
|
|
| XM_017006977.1 |
XP_016862466.1 |
1, 2
|
335
|
|
| XM_017006978.1 |
XP_016862467.1 |
2, 3
|
158
|
|
| XM_017006979.1 |
XP_016862468.1 |
1, 2
|
647
|
|
| XM_017006980.1 |
XP_016862469.1 |
1, 2
|
379
|
|
| GenBank mRNA |
AF268872.1 |
- |
1, 2
|
117
|
|
| BC007933.2 |
- |
1, 2
|
256
|
4120858 |
| BG420135.1 |
- |
1, 2
|
365
|
|
| DB454660.1 |
- |
1, 2
|
381
|
|
Back To Top
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System