Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RORB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006914.3 | NP_008845.2 | 3 | 759 | ||
GenBank mRNA | AB307695.1 | - | 2 | 118 | ||
AK313431.1 | - | 3 | 427 | |||
BC051830.1 | - | 3 | 747 | 6141868 | ||
BC093772.1 | - | 3 | 193 | 7939617 | ||
BC093774.1 | - | 3 | 193 | 7939619 | ||
BX647070.1 | - | 3 | 201 | |||
HQ709186.1 | - | 2 | 118 | |||
Y08639.1 | - | 3 | 712 |