Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BDNF SIRNA Assays ›
          Gene Symbol:
          BDNF BDNF-AS
          Gene Name:
          brain derived neurotrophic factor
          BDNF antisense RNA
          Gene Aliases:
          ANON2, BULN2
          ANTI-BDNF, BDNF, BDNF-AS1, BDNFOS, NCRNA00049
          Chromosome Location:
          Chr.11: 27654893 - 27722058 on Build GRCh38
          Chromosome Location:
          Chr.11: 27506852 - 27698171 on Build GRCh38
          UniGene ID:
          Hs.502182
          Hs.675323
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS184489
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BDNF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001143805.1 NP_001137277.1 2 315
          NM_001143806.1 NP_001137278.1 2 530
          NM_001143807.1 NP_001137279.1 2 416
          NM_001143808.1 NP_001137280.1 2 252
          NM_001143809.1 NP_001137281.1 2 262
          NM_001143810.1 NP_001137282.1 3 545
          NM_001143811.1 NP_001137283.1 4 662
          NM_001143812.1 NP_001137284.1 2 404
          NM_001143813.1 NP_001137285.1 2 486
          NM_001143814.1 NP_001137286.1 3 611
          NM_001143816.1 NP_001137288.1 2 1009
          NM_001709.4 NP_001700.2 2 504
          NM_170731.4 NP_733927.1 2 826
          NM_170732.4 NP_733928.1 2 613
          NM_170733.3 NP_733929.1 2 516
          NM_170734.3 NP_733930.1 2 464
          NM_170735.5 NP_733931.1 1 1243
          XM_011520280.2 XP_011518582.1 2 1410
          GenBank mRNA AF400438.1 - 1 176
          AK289763.1 - 2 312
          AK289853.1 - 2 213
          AY054392.2 - 2 517
          AY054393.2 - 2 259
          AY054394.2 - 2 474
          AY054395.2 - 2 557
          AY054396.2 - 2 315
          AY054397.2 - 2 398
          AY054398.2 - 2 445
          AY054399.2 - 3 330
          AY054400.2 - 2 431
          AY054406.3 - 2 299
          AY656701.1 - 1 158
          BC029795.1 - 2 489 5193877
          CX751420.1 - 2 383
          EF674517.1 - 2 315
          EF674518.1 - 2 530
          EF674519.1 - 2 613
          EF674520.1 - 2 416
          EF674521.1 - 2 516
          EF689009.1 - 1 1257
          EF689010.1 - 2 1023
          EF689011.1 - 2 252
          EF689012.1 - 2 262
          EF689013.1 - 2 404
          EF689014.1 - 2 486
          EF689015.1 - 2 504
          EF689016.1 - 3 611
          EF689017.1 - 3 363
          EF689018.1 - 2 464
          EF689019.1 - 3 545
          EF689020.1 - 4 662
          EF689021.1 - 2 826
          KP229412.1 - 1 158
          KU177943.1 - 1 158
          KU177944.1 - 1 158
          KU177945.1 - 1 158
          M61176.1 - 1 720
          X60201.1 - 2 454
          X91251.1 - 2 442
          Gene Symbol: BDNF-AS
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002832.2 5 537
          NR_033312.1 4 468
          NR_033313.1 4 468
          NR_033314.1 5 537
          NR_033315.1 4 468
          GenBank mRNA AY054401.3 - 4 468
          AY054402.3 - 4 468
          AY054404.4 - 4 468
          AY513486.3 - 4 468
          EF689023.1 - 4 473
          EF689024.1 - 3 376
          EF689025.1 - 3 376
          EF689026.1 - 3 376
          EF689027.1 - 3 376
          EF689028.1 - 3 376
          EF689029.1 - 3 376
          EF689030.1 - 5 542
          EF689031.1 - 5 542
          EF689032.1 - 5 542
          EF689033.1 - 5 542
          EF689034.1 - 5 542
          EF689035.1 - 5 542
          EF689036.1 - 4 473
          EF689037.1 - 4 473
          EF689038.1 - 4 473
          EF689039.1 - 4 473
          EF689040.1 - 4 473
          EF689041.1 - 4 473
          EF689042.1 - 5 542

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          neurotrophic factor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell-cell signaling
          nervous system development
          axon guidance
          synapse assembly
          brain-derived neurotrophic factor receptor signaling pathway
          positive regulation of brain-derived neurotrophic factor receptor signaling pathway
          negative regulation of neuron apoptotic process
          collateral sprouting
          positive regulation of collateral sprouting
          positive regulation of synapse assembly
          neurotrophin TRKB receptor binding
          growth factor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0