Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001247999.1 | NP_001234928.1 | 6 | 956 | ||
NM_022349.3 | NP_071744.2 | 5 | 767 | |||
NM_152851.2 | NP_690590.1 | 5 | 767 | |||
NM_152852.2 | NP_690591.1 | 6 | 892 | |||
XM_005274177.2 | XP_005274234.1 | 6 | 951 | |||
XM_006718658.2 | XP_006718721.1 | 6 | 826 | |||
XM_006718660.2 | XP_006718723.1 | 5 | 691 | |||
XM_006718661.2 | XP_006718724.1 | 5 | 691 | |||
XM_017018123.1 | XP_016873612.1 | 4 | 427 | |||
XM_017018124.1 | XP_016873613.1 | 4 | 427 | |||
GenBank mRNA | AB013104.1 | - | 5 | 475 | ||
AF142409.1 | - | 5 | 495 | |||
AF212240.1 | - | 5 | 636 | |||
AF237908.1 | - | 5 | 637 | |||
AF253977.1 | - | 5 | 640 | |||
AF261136.1 | - | 5 | 637 | |||
AF286866.1 | - | 4 | 415 | |||
AF350502.1 | - | 5 | 484 | |||
AF350503.1 | - | 5 | 484 | |||
AF354930.1 | - | 5 | 637 | |||
AK291870.1 | - | 6 | 892 | |||
AK300725.1 | - | 5 | 501 | |||
AK301387.1 | - | 4 | 327 | |||
BC022854.1 | - | 5 | 506 | 3934920 | ||
BC108728.1 | - | 5 | 540 | 6196213 | ||
DA979138.1 | - | 5 | 538 |