Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039763.3 | NP_001034852.3 | 10 | 1286 | ||
XM_006714670.3 | XP_006714733.1 | 12 | 1662 | |||
XM_011543552.2 | XP_011541854.1 | 13 | 2013 | |||
XM_011543553.2 | XP_011541855.1 | 10 | 2096 | |||
XM_011543555.2 | XP_011541857.1 | 12 | 1877 | |||
XM_011543556.1 | XP_011541858.1 | 10 | 1607 | |||
XM_011543557.2 | XP_011541859.1 | 13 | 2014 | |||
XM_011543559.2 | XP_011541861.1 | 13 | 2013 | |||
XM_011543560.2 | XP_011541862.1 | 8 | 986 | |||
XM_011543561.2 | XP_011541863.1 | 13 | 2013 | |||
XM_011543563.2 | XP_011541865.1 | 13 | 2012 | |||
XM_011543564.2 | XP_011541866.1 | 13 | 2011 | |||
XM_011543565.2 | XP_011541867.1 | 13 | 2011 | |||
XM_011543566.2 | XP_011541868.1 | 10 | 1368 | |||
XM_011543567.2 | XP_011541869.1 | 13 | 2011 | |||
XM_017009704.1 | XP_016865193.1 | 11 | 2242 | |||
XM_017009705.1 | XP_016865194.1 | 13 | 2014 | |||
XM_017009706.1 | XP_016865195.1 | 13 | 2013 | |||
XM_017009707.1 | XP_016865196.1 | 11 | 1505 | |||
GenBank mRNA | AK125070.1 | - | 4 | 714 | ||
AK296542.1 | - | 10 | 1202 | |||
BC042563.1 | - | 9 | 1069 | |||
BX648527.1 | - | 8 | 932 |