Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048249.3 | NP_001041714.1 | 3 | 495 | ||
GenBank mRNA | AK291180.1 | - | 4 | 404 | ||
BC014242.2 | - | 3 | 349 | |||
BC047489.2 | - | 3 | 376 | |||
BC066550.1 | - | 3 | 474 | |||
BC103711.1 | - | 3 | 450 | |||
BC146876.1 | - | 3 | 340 | 9056804 | ||
BC146881.1 | - | 3 | 340 | 9056809 | ||
BX538015.1 | - | 3 | 490 | |||
BX648459.1 | - | 3 | 467 | |||
DA039277.1 | - | 3 | 495 |