Search
Search

TXNRD3NB| Gene Symbol: TXNRD3NB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039783.1 | NP_001034872.1 | 3 | 510 | ||
| GenBank mRNA | AB113648.1 | - | 3 | 510 | ||
| BC130546.1 | - | 1 | 124 | 40146577 | ||
| BC130548.1 | - | 1 | 124 | 40146579 | ||