Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101404.1 | NP_001094874.1 | 5 | 1285 | ||
XM_011521895.2 | XP_011520197.1 | 5 | 2577 | |||
XM_011521896.2 | XP_011520198.1 | 6 | 1261 | |||
XM_011521899.2 | XP_011520201.1 | 4 | 2052 | |||
XM_017022487.1 | XP_016877976.1 | 6 | 1494 | |||
XM_017022488.1 | XP_016877977.1 | 4 | 1069 |