Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EFCAB6 SIRNA Assays ›
Gene Symbol:
EFCAB6
Gene Name:
EF-hand calcium binding domain 6
Gene Aliases:
DJBP, HSCBCIP1, dJ185D5.1
Chromosome Location:
Chr.22: 43528549 - 43812403 on Build GRCh38
UniGene ID:
Hs.658996
Species:
Human
siRNA ID
HSS185578
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EFCAB6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022785.3
NP_073622.2
25
3366
NM_198856.2
NP_942153.1
23
3008
XM_005261704.2
XP_005261761.1
25
3306
XM_005261705.3
XP_005261762.1
24
3220
XM_011530316.1
XP_011528618.1
26
3593
XM_011530317.2
XP_011528619.1
24
3153
XM_011530318.2
XP_011528620.1
24
3249
XM_011530319.2
XP_011528621.1
23
2941
XM_011530320.2
XP_011528622.1
26
3405
XM_011530321.2
XP_011528623.1
23
2936
XM_011530322.2
XP_011528624.1
24
3064
XM_011530323.2
XP_011528625.1
21
2722
XM_011530325.2
XP_011528627.1
19
2467
XM_011530326.2
XP_011528628.1
26
3405
XM_011530327.2
XP_011528629.1
16
2214
XM_011530329.1
XP_011528631.1
12
1686
XM_017028910.1
XP_016884399.1
26
3405
XM_017028911.1
XP_016884400.1
26
3405
XM_017028912.1
XP_016884401.1
22
3064
GenBank mRNA
AB051459.1
-
11
1549
AB073862.1
-
5
560
AK027241.1
-
5
564
AK225596.1
-
5
565
AK225605.1
-
5
565
AK292411.1
-
25
3306
AK292498.1
-
23
2985
AK309013.1
-
6
764
BC039315.1
-
23
2861
5297436
Back To Top
More Information
Panther Classification:
Molecular Function -
calmodulin-related
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0