Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022789.3 | NP_073626.1 | 1 | 332 | ||
NM_172314.1 | NP_758525.1 | 2 | 184 | |||
GenBank mRNA | AF305200.1 | - | 1 | 332 | ||
AF458059.1 | - | 1 | 26 | |||
AF461739.1 | - | 2 | 184 | |||
AY359127.1 | - | 1 | 332 | |||
BC069565.1 | - | 1 | 149 | 7262228 | ||
BC104929.1 | - | 2 | 74 | 8143932 | ||
BC104931.1 | - | 2 | 74 | 8143934 |