Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SKI SIRNA Assays ›
Gene Symbol:
SKI
Gene Name:
SKI proto-oncogene
Gene Aliases:
SGS, SKV
Chromosome Location:
Chr.1: 2228695 - 2310213 on Build GRCh38
UniGene ID:
Hs.656507
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS185683
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SKI
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003036.3
NP_003027.1
1
149
XM_005244775.3
XP_005244832.1
1
119
GenBank mRNA
X15218.1
-
1
149
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008764420.
This siRNA also targets the following transcripts in Mouse: NM_011385, BC068305, AF435852.
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
neural tube closure
lens morphogenesis in camera-type eye
transcription, DNA-templated
transforming growth factor beta receptor signaling pathway
cell proliferation
negative regulation of cell proliferation
anterior/posterior axis specification
negative regulation of Schwann cell proliferation
myotube differentiation
olfactory bulb development
myelination in peripheral nervous system
positive regulation of Wnt signaling pathway
embryonic limb morphogenesis
BMP signaling pathway
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of BMP signaling pathway
negative regulation of histone deacetylation
negative regulation of activin receptor signaling pathway
somatic stem cell population maintenance
regulation of apoptotic process
camera-type eye development
positive regulation of DNA binding
nose morphogenesis
negative regulation of osteoblast differentiation
positive regulation of transcription from RNA polymerase II promoter
negative regulation of fibroblast proliferation
camera-type eye morphogenesis
skeletal muscle fiber development
cell motility
palate development
retina development in camera-type eye
face morphogenesis
bone morphogenesis
SMAD protein signal transduction
protein homotrimerization
transcription corepressor activity
protein binding
zinc ion binding
protein kinase binding
protein domain specific binding
ubiquitin protein ligase binding
SMAD binding
histone deacetylase inhibitor activity
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-dwbsp:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0