Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NDRG4 SIRNA Assays ›
Gene Symbol:
NDRG4
Gene Name:
NDRG family member 4
Gene Aliases:
BDM1, SMAP-8, SMAP8
Chromosome Location:
Chr.16: 58463645 - 58515403 on Build GRCh38
UniGene ID:
Hs.322430
Species:
Human
siRNA ID
HSS185700
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NDRG4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130487.1
NP_001123959.1
13
1261
NM_001242833.1
NP_001229762.1
12
960
NM_001242834.1
NP_001229763.1
11
924
NM_001242835.1
NP_001229764.1
11
870
NM_001242836.1
NP_001229765.1
11
870
NM_020465.3
NP_065198.1
13
1036
NM_022910.3
NP_075061.1
13
1133
NR_040072.1
11
962
XM_006721252.3
XP_006721315.1
14
1136
XM_006721253.3
XP_006721316.1
14
1069
XM_006721254.3
XP_006721317.1
13
1123
XM_006721256.3
XP_006721319.1
14
1067
XM_006721257.3
XP_006721320.1
13
1108
XM_006721258.3
XP_006721321.1
13
1012
XM_006721259.3
XP_006721322.1
13
1077
XM_006721260.3
XP_006721323.1
13
1015
XM_006721261.3
XP_006721324.1
12
960
XM_006721262.3
XP_006721325.1
11
1041
XM_011523290.2
XP_011521592.1
14
1223
XM_011523291.2
XP_011521593.1
13
1187
XM_011523292.2
XP_011521594.1
14
1223
XM_011523293.2
XP_011521595.1
13
1181
XM_011523294.2
XP_011521596.1
14
1002
XM_017023582.1
XP_016879071.1
13
1187
XM_017023583.1
XP_016879072.1
14
1223
XM_017023584.1
XP_016879073.1
14
1136
XM_017023585.1
XP_016879074.1
13
1123
XM_017023586.1
XP_016879075.1
13
1070
XM_017023587.1
XP_016879076.1
14
1067
XM_017023588.1
XP_016879077.1
13
1012
XM_017023589.1
XP_016879078.1
13
1077
XM_017023590.1
XP_016879079.1
14
1002
GenBank mRNA
AB044944.1
-
11
870
AB044945.1
-
11
870
AB044946.1
-
12
883
AK126574.1
-
13
1033
AK126729.1
-
14
2983
AK131248.1
-
11
924
AK296415.1
-
11
924
AL136584.1
-
11
856
BC001527.1
-
11
854
BC011795.2
-
11
858
4139594
KU178760.1
-
11
764
KU178761.1
-
11
764
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
heart looping
response to stress
brain development
visual learning
negative regulation of platelet-derived growth factor receptor signaling pathway
positive regulation of neuron projection development
negative regulation of smooth muscle cell migration
cell growth
cell differentiation
embryonic heart tube development
vesicle docking
negative regulation of smooth muscle cell proliferation
cardiac muscle cell proliferation
cell migration involved in heart development
positive regulation of ERK1 and ERK2 cascade
regulation of endocytic recycling
molecular_function
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-zbtnl:80/10.239.166.103:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0