Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003098.2 | NP_003089.1 | 6, 7 | 1507 | ||
XM_005260517.1 | XP_005260574.1 | 6, 7 | 1468 | |||
GenBank mRNA | AK095942.1 | - | 6, 7 | 1067 | ||
AK291994.1 | - | 6, 7 | 1378 | |||
AK301800.1 | - | 6, 7 | 1013 | |||
BC026215.2 | - | 6, 7 | 1347 | 4413197 | ||
S81737.1 | - | 6, 7 | 1367 | |||
U40571.1 | - | 6, 7 | 1272 |