Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DST SIRNA Assays ›
Gene Symbol:
DST
Gene Name:
dystonin
Gene Aliases:
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
Chromosome Location:
Chr.6: 56457987 - 56954671 on Build GRCh38
UniGene ID:
Hs.604915
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS186057
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DST
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144769.2
NP_001138241.1
34
4743
NM_001144770.1
NP_001138242.1
32
4414
NM_001723.5
NP_001714.1
21
3298
NM_015548.4
NP_056363.2
21
3298
NM_183380.3
NP_899236.1
31
4688
XM_005249310.3
XP_005249367.1
35
4837
XM_005249315.3
XP_005249372.1
34
4784
XM_005249316.2
XP_005249373.1
27
5060
XM_005249318.2
XP_005249375.1
33
4498
XM_005249319.2
XP_005249376.1
32
4399
XM_005249320.4
XP_005249377.1
32
4455
XM_005249322.4
XP_005249379.1
32
4350
XM_005249323.3
XP_005249380.1
31
4683
XM_005249324.2
XP_005249381.1
21
3295
XM_011514824.2
XP_011513126.1
34
4748
XM_011514825.2
XP_011513127.1
34
4744
XM_011514826.2
XP_011513128.1
31
4683
XM_017011205.1
XP_016866694.1
35
4847
XM_017011206.1
XP_016866695.1
35
4847
XM_017011207.1
XP_016866696.1
35
4784
XM_017011208.1
XP_016866697.1
35
4847
XM_017011209.1
XP_016866698.1
35
4847
XM_017011210.1
XP_016866699.1
35
4847
XM_017011211.1
XP_016866700.1
35
4847
XM_017011212.1
XP_016866701.1
35
4847
XM_017011213.1
XP_016866702.1
35
4847
XM_017011214.1
XP_016866703.1
35
4847
XM_017011215.1
XP_016866704.1
35
4847
XM_017011216.1
XP_016866705.1
35
4847
XM_017011217.1
XP_016866706.1
32
4428
XM_017011219.1
XP_016866708.1
35
4843
XM_017011220.1
XP_016866709.1
34
4786
XM_017011221.1
XP_016866710.1
35
4843
XM_017011222.1
XP_016866711.1
27
5002
XM_017011223.1
XP_016866712.1
35
4843
XM_017011224.1
XP_016866713.1
31
4683
XM_017011225.1
XP_016866714.1
35
4857
GenBank mRNA
AF400226.1
-
21
3190
AF400227.1
-
21
3190
AK299898.1
-
32
4414
BC065536.1
-
21
3262
L11690.1
-
21
3292
M63618.1
-
11
1529
M69225.1
-
21
3228
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
cell adhesion
integrin-mediated signaling pathway
response to wounding
maintenance of cell polarity
intermediate filament cytoskeleton organization
cell motility
actin binding
integrin binding
calcium ion binding
protein binding
protein C-terminus binding
microtubule plus-end binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0