Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000582.2 | NP_000573.1 | 6 | 757 | ||
NM_001040058.1 | NP_001035147.1 | 7 | 799 | |||
NM_001040060.1 | NP_001035149.1 | 6 | 718 | |||
NM_001251829.1 | NP_001238758.1 | 5 | 676 | |||
NM_001251830.1 | NP_001238759.1 | 8 | 988 | |||
XM_017008564.1 | XP_016864053.1 | 5 | 1225 | |||
GenBank mRNA | AB209987.1 | - | 4 | 511 | ||
AB469789.1 | - | 4 | 511 | |||
AF052124.1 | - | 6 | 679 | |||
AK057738.1 | - | 7 | 735 | |||
AK075463.1 | - | 8 | 669 | |||
AK290090.1 | - | 6 | 654 | |||
AK290104.1 | - | 7 | 736 | |||
AK295491.1 | - | 8 | 923 | |||
AK296035.1 | - | 6 | 654 | |||
AK315461.1 | - | 6 | 687 | |||
BC007016.1 | - | 6 | 693 | 4052438 | ||
BC017387.1 | - | 7 | 739 | 3828885 | ||
BC022844.1 | - | 6 | 694 | 4284921 | ||
BC093033.1 | - | 6 | 710 | 5288027 | ||
BX648003.1 | - | 6 | 672 | |||
D28759.1 | - | 6 | 634 | |||
D28760.1 | - | 5 | 592 | |||
D28761.1 | - | 5 | 553 | |||
DQ839491.1 | - | 6 | 634 | |||
DQ846870.1 | - | 6 | 634 | |||
DQ846871.1 | - | 6 | 634 | |||
J04765.1 | - | 6 | 700 | |||
JF412666.1 | - | 7 | 570 | |||
JF412667.1 | - | 6 | 716 | |||
KR062183.1 | - | 8 | 923 | |||
M83248.1 | - | 7 | 731 | |||
X13694.1 | - | 7 | 701 |