Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TCF12 SIRNA Assays ›
          Gene Symbol:
          TCF12
          Gene Name:
          transcription factor 12
          Gene Aliases:
          CRS3, HEB, HTF4, HsT17266, TCF-12, bHLHb20
          Chromosome Location:
          Chr.15: 56918090 - 57291261 on Build GRCh38
          UniGene ID:
          Hs.511504
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS186237
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TCF12
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001306219.2 NP_001293148.1 5, 6 609
          NM_001306220.2 NP_001293149.1 4, 5 469
          NM_001322151.1 NP_001309080.1 12, 13 1316
          NM_001322152.1 NP_001309081.1 12, 13 1321
          NM_001322154.1 NP_001309083.1 12, 13 1316
          NM_001322156.1 NP_001309085.1 13, 14 1383
          NM_001322157.1 NP_001309086.1 12, 13 1321
          NM_001322158.1 NP_001309087.1 13, 14 1383
          NM_001322159.1 NP_001309088.1 12, 13 1321
          NM_001322161.1 NP_001309090.1 12, 13 1167
          NM_001322162.1 NP_001309091.1 12, 13 1167
          NM_001322164.1 NP_001309093.1 12, 13 1186
          NM_001322165.1 NP_001309094.1 11, 12 1150
          NM_003205.3 NP_003196.1 12, 13 1321
          NM_207036.1 NP_996919.1 12, 13 1311
          NM_207037.1 NP_996920.1 12, 13 1321
          NM_207038.1 NP_996921.1 12, 13 1311
          NM_207040.1 NP_996923.1 5, 6 582
          XM_011521959.2 XP_011520261.1 13, 14 1317
          XM_011521960.2 XP_011520262.1 13, 14 1317
          XM_011521961.2 XP_011520263.1 13, 14 1317
          XM_011521962.2 XP_011520264.1 13, 14 1317
          XM_011521963.2 XP_011520265.1 13, 14 1317
          XM_011521965.1 XP_011520267.1 11, 12 1028
          XM_011521966.2 XP_011520268.1 4, 5 924
          XM_011521967.1 XP_011520269.1 11, 12 1028
          XM_011521969.1 XP_011520271.1 5, 6 575
          XM_017022520.1 XP_016878009.1 12, 13 1287
          GenBank mRNA AK294617.1 - 12, 13 1269
          AK294991.1 - 5, 6 609
          AK302923.1 - 12, 13 1271
          AK304007.1 - 4, 5 464
          AK308101.1 - 5, 6 583
          AK311359.1 - 5, 6 603
          AK312710.1 - 12, 13 1281
          AL831981.2 - 12, 13 1289
          AU120213.1 - 5, 6 603
          BC050556.1 - 12, 13 1262 5767579
          BX537967.1 - 5, 6 573
          M80627.1 - 12, 13 1240
          M83233.1 - 12, 13 1115

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          immune response
          muscle organ development
          positive regulation of neuron differentiation
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          enhancer binding
          cAMP response element binding
          protein homodimerization activity
          bHLH transcription factor binding
          transcription regulatory region DNA binding
          SMAD binding
          protein heterodimerization activity
          E-box binding
          HMG box domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0