Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099691.2 | NP_001093161.1 | 5 | 700 | ||
NM_001308158.1 | NP_001295087.1 | 5 | 870 | |||
NM_001308159.1 | NP_001295088.1 | 5 | 867 | |||
NM_003236.3 | NP_003227.1 | 5 | 703 | |||
GenBank mRNA | AF149096.1 | - | 5 | 460 | ||
AF149097.1 | - | 5 | 457 | |||
AF149098.1 | - | 5 | 457 | |||
AK290151.1 | - | 5 | 514 | |||
AK309066.1 | - | 5 | 870 | |||
AK309220.1 | - | 5 | 719 | |||
AK312899.1 | - | 5 | 511 | |||
BC005308.1 | - | 5 | 511 | 3828239 | ||
BT006833.1 | - | 5 | 452 | |||
K03222.1 | - | 5 | 489 | |||
M31172.1 | - | 5 | 489 | |||
X70340.1 | - | 5 | 486 |