Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TLE1 SIRNA Assays ›
Gene Symbol:
TLE1
Gene Name:
transducin like enhancer of split 1
Gene Aliases:
ESG, ESG1, GRG1
Chromosome Location:
Chr.9: 81583683 - 81689535 on Build GRCh38
UniGene ID:
Hs.197320
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS186345
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TLE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001303103.1
NP_001290032.1
9
1948
NM_001303104.1
NP_001290033.1
9
1918
NM_005077.4
NP_005068.2
9
1918
XM_005252151.1
XP_005252208.1
9
862
XM_005252152.1
XP_005252209.1
9
862
XM_005252153.1
XP_005252210.1
9
832
XM_005252154.1
XP_005252211.1
9
829
XM_005252156.2
XP_005252213.1
9
862
XM_005252162.1
XP_005252219.1
7
610
XM_005252163.2
XP_005252220.1
7
610
XM_006717258.1
XP_006717321.1
9
859
XM_006717259.3
XP_006717322.1
7
545
XM_006717260.1
XP_006717323.1
7
640
XM_006717261.2
XP_006717324.1
7
574
XM_006717262.1
XP_006717325.1
9
832
XM_006717263.1
XP_006717326.1
9
829
XM_011518951.2
XP_011517253.1
9
829
XM_017015064.1
XP_016870553.1
9
859
XM_017015065.1
XP_016870554.1
9
859
XM_017015066.1
XP_016870555.1
7
602
GenBank mRNA
AB209854.1
-
9
1918
AK290860.1
-
9
680
AK293606.1
-
9
706
AK304565.1
-
7
519
BC010100.2
-
9
1064
3506451
BC015747.1
-
9
774
4857394
DB211613.1
-
7
491
M99435.1
-
9
648
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
signal transduction
multicellular organism development
organ morphogenesis
positive regulation of gene expression
Wnt signaling pathway
negative regulation of Wnt signaling pathway
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of transcription, DNA-templated
beta-catenin-TCF complex assembly
negative regulation of anoikis
RNA polymerase II transcription corepressor activity
protein binding
transcription factor binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0