Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TP53 SIRNA Assays ›
          Gene Symbol:
          TP53
          Gene Name:
          tumor protein p53
          Gene Aliases:
          BCC7, LFS1, P53, TRP53
          Chromosome Location:
          Chr.17: 7668402 - 7687550 on Build GRCh38
          UniGene ID:
          Hs.437460
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS186391
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TP53
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000546.5 NP_000537.3 6, 7 876
          NM_001126112.2 NP_001119584.1 6, 7 873
          NM_001126113.2 NP_001119585.1 6, 7 876
          NM_001126114.2 NP_001119586.1 6, 7 876
          NM_001126115.1 NP_001119587.1 2, 3 556
          NM_001126116.1 NP_001119588.1 2, 3 556
          NM_001126117.1 NP_001119589.1 2, 3 556
          NM_001126118.1 NP_001119590.1 5, 6 993
          NM_001276695.1 NP_001263624.1 6, 7 876
          NM_001276696.1 NP_001263625.1 6, 7 876
          NM_001276697.1 NP_001263626.1 2, 3 556
          NM_001276698.1 NP_001263627.1 2, 3 556
          NM_001276699.1 NP_001263628.1 2, 3 556
          NM_001276760.1 NP_001263689.1 6, 7 876
          NM_001276761.1 NP_001263690.1 6, 7 873
          GenBank mRNA AB082923.1 - 6, 7 737
          AF307851.1 - 6, 7 809
          AK225838.1 - 6, 7 814
          AK297462.1 - 7, 8 741
          AK297927.1 - 7, 8 786
          AK312568.1 - 6, 7 818
          AM076970.1 - 5, 6 641
          AM076971.1 - 5, 6 674
          AM076972.1 - 5, 6 674
          AY429684.1 - 5, 6 674
          BC003596.1 - 6, 7 794 3544714
          BT019622.1 - 5, 6 674
          DQ186648.1 - 6, 7 806
          DQ186649.1 - 6, 7 806
          DQ186650.1 - 2, 3 552
          DQ186651.1 - 2, 3 552
          DQ186652.1 - 2, 3 552
          DQ191317.1 - 6, 7 863
          DQ263704.1 - 5, 6 674
          DQ286964.1 - 6, 7 808
          DQ648884.1 - 5, 6 474
          DQ648885.1 - 3, 4 350
          EF101867.1 - 5, 6 641
          EF101868.1 - 5, 6 674
          EF101869.1 - 5, 6 674
          FJ207420.1 - 6, 7 809
          JN214348.1 - 5, 6 719
          JN601513.1 - 5, 6 674
          JN601514.1 - 5, 6 674
          JN601515.1 - 5, 6 674
          JN601516.1 - 6, 7 648
          JN900492.1 - 5, 6 982
          JQ026207.1 - 5, 6 664
          JQ026208.1 - 5, 6 669
          JQ026210.1 - 5, 6 674
          JQ694049.1 - 5, 6 697
          JQ694050.1 - 5, 6 697
          JQ694051.1 - 5, 6 697
          KC820708.1 - 2, 3 169
          KC820709.1 - 2, 3 169
          KC820710.1 - 2, 3 169
          KC820711.1 - 2, 3 169
          KC820712.1 - 2, 3 169
          KC820713.1 - 2, 3 169
          KC820714.1 - 2, 3 169
          KC820715.1 - 2, 3 169
          KC820716.1 - 2, 3 169
          KC820717.1 - 2, 3 169
          KC820718.1 - 2, 3 169
          KC820719.1 - 2, 3 169
          KC820720.1 - 2, 3 169
          KC820721.1 - 2, 3 169
          KC820722.1 - 2, 3 169
          KC820723.1 - 2, 3 169
          KC820724.1 - 2, 3 169
          KC820725.1 - 2, 3 169
          KC820726.1 - 2, 3 169
          KC820727.1 - 2, 3 169
          KC820728.1 - 2, 3 169
          KC820729.1 - 2, 3 169
          KC820730.1 - 2, 3 169
          KC820731.1 - 2, 3 169
          KC820732.1 - 2, 3 169
          KC820733.1 - 2, 3 169
          KC820734.1 - 2, 3 169
          KC820735.1 - 2, 3 169
          KC820736.1 - 2, 3 169
          KC820737.1 - 2, 3 169
          KC820738.1 - 2, 3 169
          KC820739.1 - 2, 3 169
          KC820740.1 - 2, 3 169
          KC820741.1 - 2, 3 169
          KC820742.1 - 2, 3 169
          KC820743.1 - 2, 3 169
          KC820744.1 - 2, 3 169
          KC820745.1 - 2, 3 169
          KC820746.1 - 2, 3 169
          KC820747.1 - 2, 3 169
          KC820748.1 - 2, 3 169
          KC820749.1 - 2, 3 169
          KC820750.1 - 2, 3 169
          KC820751.1 - 2, 3 169
          KC820752.1 - 2, 3 169
          KC820753.1 - 2, 3 169
          KC820754.1 - 2, 3 169
          KC820755.1 - 2, 3 169
          KC820756.1 - 2, 3 169
          KC820757.1 - 2, 3 169
          KC820758.1 - 2, 3 169
          KC820760.1 - 2, 3 169
          KC820761.1 - 2, 3 169
          KC820762.1 - 2, 3 169
          KC820763.1 - 2, 3 169
          KC820764.1 - 2, 3 169
          KC820765.1 - 2, 3 169
          KC820766.1 - 2, 3 169
          KC820767.1 - 2, 3 169
          KC820768.1 - 2, 3 169
          KC820769.1 - 2, 3 169
          KC820770.1 - 2, 3 169
          KC820771.1 - 2, 3 169
          KC820772.1 - 2, 3 169
          KC820773.1 - 2, 3 169
          KC820774.1 - 2, 3 169
          KC820775.1 - 2, 3 169
          KC820776.1 - 2, 3 169
          KC820777.1 - 2, 3 169
          KC820778.1 - 2, 3 169
          KC820779.1 - 2, 3 169
          KC820780.1 - 2, 3 169
          KC820781.1 - 2, 3 169
          KC820782.1 - 2, 3 169
          KC820783.1 - 2, 3 169
          KC820784.1 - 2, 3 169
          KC820785.1 - 2, 3 169
          KC820786.1 - 2, 3 169
          M14694.1 - 6, 7 799
          M14695.1 - 6, 7 795
          X01405.1 - 3, 4 375
          X02469.1 - 6, 7 809
          X60010.1 - 5, 6 673
          X60011.1 - 5, 6 674
          X60012.1 - 5, 6 674
          X60013.1 - 5, 6 674
          X60014.1 - 5, 6 674
          X60015.1 - 5, 6 674
          X60016.1 - 5, 6 674
          X60017.1 - 5, 6 674
          X60018.1 - 5, 6 674
          X60019.1 - 5, 6 674
          X60020.1 - 5, 6 674

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: XM_003749209.

          Panther Classification:

          Molecular Function -

          P53-like transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          DNA strand renaturation
          base-excision repair
          nucleotide-excision repair
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          protein complex assembly
          cellular response to DNA damage stimulus
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
          ER overload response
          cell cycle arrest
          Ras protein signal transduction
          multicellular organism development
          cell aging
          protein localization
          cell proliferation
          negative regulation of cell proliferation
          determination of adult lifespan
          response to X-ray
          response to gamma radiation
          positive regulation of gene expression
          viral process
          protein sumoylation
          cell differentiation
          negative regulation of cell growth
          DNA damage response, signal transduction by p53 class mediator
          positive regulation of histone deacetylation
          chromatin assembly
          mitotic G1 DNA damage checkpoint
          positive regulation of protein oligomerization
          cellular response to UV
          cellular response to drug
          cellular response to glucose starvation
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          regulation of apoptotic process
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          entrainment of circadian clock by photoperiod
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of neuron apoptotic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          response to antibiotic
          positive regulation of protein export from nucleus
          regulation of mitochondrial membrane permeability
          negative regulation of fibroblast proliferation
          circadian behavior
          positive regulation of peptidyl-tyrosine phosphorylation
          negative regulation of helicase activity
          protein tetramerization
          negative regulation of telomerase activity
          positive regulation of thymocyte apoptotic process
          positive regulation of cell cycle arrest
          cellular response to hypoxia
          cellular response to ionizing radiation
          intrinsic apoptotic signaling pathway by p53 class mediator
          positive regulation of release of cytochrome c from mitochondria
          replicative senescence
          oxidative stress-induced premature senescence
          intrinsic apoptotic signaling pathway
          oligodendrocyte apoptotic process
          positive regulation of execution phase of apoptosis
          positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
          regulation of signal transduction by p53 class mediator
          regulation of cell cycle G2/M phase transition
          positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
          positive regulation of reactive oxygen species metabolic process
          positive regulation of intrinsic apoptotic signaling pathway
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          RNA polymerase II transcription factor binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          protease binding
          p53 binding
          DNA binding
          chromatin binding
          damaged DNA binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          copper ion binding
          protein binding
          ATP binding
          transcription factor binding
          zinc ion binding
          enzyme binding
          protein kinase binding
          protein phosphatase binding
          receptor tyrosine kinase binding
          ubiquitin protein ligase binding
          histone acetyltransferase binding
          identical protein binding
          sequence-specific DNA binding
          protein self-association
          transcription regulatory region DNA binding
          protein heterodimerization activity
          protein N-terminus binding
          chaperone binding
          protein phosphatase 2A binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0