Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007344.3 | NP_031370.2 | 2 | 504 | ||
NR_134525.1 | 2 | 504 | ||||
XM_006717273.3 | XP_006717336.2 | 2 | 512 | |||
XM_017015105.1 | XP_016870594.1 | 2 | 1011 | |||
GenBank mRNA | AK225119.1 | - | 2 | 490 | ||
AK302902.1 | - | 2 | 490 | |||
BC050734.1 | - | 1 | 466 | |||
BC062692.1 | - | 2 | 488 | |||
BC104639.1 | - | 2 | 488 | |||
BC143821.1 | - | 2 | 477 | |||
BM827593.1 | - | 2 | 487 | |||
BP209178.1 | - | 2 | 504 |