Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034176.1 | 4 | 745 | |||
XM_011511014.2 | XP_011509316.1 | 3 | 851 | |||
XM_017003895.1 | XP_016859384.1 | 3 | 851 | |||
XM_017003896.1 | XP_016859385.1 | 3 | 851 | |||
XM_017003897.1 | XP_016859386.1 | 3 | 851 | |||
GenBank mRNA | AK128393.1 | - | 4 | 691 |