Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003374.2 | NP_003365.1 | 4 | 413 | ||
NR_036624.1 | 4 | 491 | ||||
NR_036625.1 | 4 | 494 | ||||
XM_005272075.3 | XP_005272132.1 | 4 | 410 | |||
XM_017009821.1 | XP_016865310.1 | 4 | 341 | |||
XM_017009822.1 | XP_016865311.1 | 4 | 320 | |||
XM_017009823.1 | XP_016865312.1 | 4 | 343 | |||
GenBank mRNA | AK095989.1 | - | 4 | 413 | ||
AK122953.1 | - | 7 | 1816 | |||
BC008482.1 | - | 4 | 199 | 4332199 | ||
BC071168.1 | - | 4 | 230 | 6023095 | ||
BC090042.1 | - | 4 | 223 | 30395350 | ||
DA072915.1 | - | 4 | 307 | |||
L06132.1 | - | 4 | 267 |