Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278284.1 | NP_001265213.1 | 3 | 459 | ||
NM_001278287.1 | NP_001265216.1 | 3 | 375 | |||
NM_001278290.1 | NP_001265219.1 | 2 | 220 | |||
NM_001318135.1 | NP_001305064.1 | 3 | 337 | |||
NM_017715.3 | NP_060185.2 | 3 | 341 | |||
NM_032924.4 | NP_116313.3 | 3 | 341 | |||
XM_006716111.1 | XP_006716174.1 | 3 | 321 | |||
XM_017012582.1 | XP_016868071.1 | 3 | 335 | |||
GenBank mRNA | AF193057.1 | - | 3 | 304 | ||
AF217988.1 | - | 3 | 305 | |||
AK000223.1 | - | 1 | 76 | |||
AK297625.1 | - | 3 | 303 | |||
AK309719.1 | - | 3 | 647 | |||
BC011887.2 | - | 3 | 265 | 3831842 | ||
BC013603.1 | - | 3 | 284 | 3865426 | ||
BC025265.1 | - | 3 | 274 | |||
BQ071498.1 | - | 3 | 171 | |||
BX647182.1 | - | 2 | 982 | |||
DC347416.1 | - | 3 | 341 | |||
DC384519.1 | - | 3 | 341 | |||
HY096067.1 | - | 2 | 199 | |||
X60153.1 | - | 1 | 14 |