Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256279.1 | NP_001243208.1 | 5 | 884 | ||
NM_001256280.1 | NP_001243209.1 | 3 | 689 | |||
NM_019591.3 | NP_062537.2 | 4 | 785 | |||
XM_005266182.3 | XP_005266239.1 | 5 | 872 | |||
XM_005266183.3 | XP_005266240.1 | 6 | 1075 | |||
XM_011534829.1 | XP_011533131.1 | 5 | 988 | |||
XM_017019920.1 | XP_016875409.1 | 6 | 1508 | |||
XM_017019921.1 | XP_016875410.1 | 6 | 971 | |||
XM_017019922.1 | XP_016875411.1 | 5 | 1857 | |||
XM_017019923.1 | XP_016875412.1 | 5 | 1788 | |||
GenBank mRNA | AK122941.1 | - | 3 | 2898 | ||
AK291243.1 | - | 4 | 752 | |||
BC046206.2 | - | 4 | 686 | 6176427 | ||
CN268178.1 | - | 3 | 594 | |||
DA737586.1 | - | 4 | 731 | |||
EU668356.1 | - | 4 | 663 |