Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DDR1 SIRNA Assays ›
Gene Symbol:
DDR1
Gene Name:
discoidin domain receptor tyrosine kinase 1
Gene Aliases:
CAK, CD167, DDR, EDDR1, HGK2, MCK10, NEP, NTRK4, PTK3, PTK3A, RTK6, TRKE
Chromosome Location:
Chr.6: 30880909 - 30900156 on Build GRCh38
UniGene ID:
Hs.631988
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS187878
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DDR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001202521.1
NP_001189450.1
6, 7
902
NM_001202522.1
NP_001189451.1
6, 7
902
NM_001202523.1
NP_001189452.1
7, 8
1154
NM_001297652.1
NP_001284581.1
7, 8
1273
NM_001297653.1
NP_001284582.1
7, 8
1075
NM_001297654.1
NP_001284583.1
7, 8
1075
NM_001954.4
NP_001945.3
9, 10
1193
NM_013993.2
NP_054699.2
8, 9
1119
NM_013994.2
NP_054700.2
6, 7
902
XM_006715185.2
XP_006715248.1
7, 8
1066
XM_011514882.2
XP_011513184.1
7, 8
1066
XM_011514883.1
XP_011513185.1
9, 10
1181
XM_011514884.1
XP_011513186.1
9, 10
1257
XM_011514885.1
XP_011513187.1
7, 8
1104
XM_011514886.1
XP_011513188.1
7, 8
969
XM_011514887.1
XP_011513189.1
7, 8
1119
XM_011514888.2
XP_011513190.1
7, 8
1066
XM_017011268.1
XP_016866757.1
7, 8
936
XM_017011269.1
XP_016866758.1
7, 8
1119
GenBank mRNA
AB210021.1
-
8, 9
1014
AK130776.1
-
2, 3
248
AK291621.1
-
7, 8
968
AK294793.1
-
7, 8
1140
BC008716.2
-
9, 10
1193
2964574
BC013400.2
-
9, 10
1193
2964574
BC070070.1
-
7, 8
964
5262509
L11315.1
-
8, 9
1001
L20817.1
-
7, 8
1052
L57508.1
-
7, 8
1052
X74979.1
-
6, 7
905
Z29093.1
-
6, 7
1196
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_013137, NM_001166022, BC166604.
This siRNA also targets the following transcripts in Mouse: NM_007584, NM_001198833, NM_001198831, NM_172962, AK138698, and others.
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
regulation of cell-matrix adhesion
cell adhesion
embryo implantation
lactation
negative regulation of cell proliferation
regulation of extracellular matrix disassembly
smooth muscle cell migration
extracellular matrix organization
organ regeneration
collagen-activated tyrosine kinase receptor signaling pathway
peptidyl-tyrosine autophosphorylation
ear development
skin development
wound healing, spreading of cells
protein autophosphorylation
branching involved in mammary gland duct morphogenesis
mammary gland alveolus development
smooth muscle cell-matrix adhesion
transmembrane receptor protein tyrosine kinase activity
protein binding
collagen binding
ATP binding
protein tyrosine kinase collagen receptor activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0