Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004633.3 | NP_004624.1 | 9 | 1305 | ||
NR_048564.1 | 9 | 1292 | ||||
XM_006712734.3 | XP_006712797.1 | 9 | 1329 | |||
XM_006712736.3 | XP_006712799.1 | 9 | 1366 | |||
XM_011511801.2 | XP_011510103.1 | 9 | 3665 | |||
XM_011511803.2 | XP_011510105.1 | 9 | 3244 | |||
XM_011511804.2 | XP_011510106.1 | 9 | 1358 | |||
XM_011511805.2 | XP_011510107.1 | 9 | 2131 | |||
XM_011511808.2 | XP_011510110.1 | 5 | 960 | |||
XM_017004889.1 | XP_016860378.1 | 8 | 1215 | |||
GenBank mRNA | AK130412.1 | - | 9 | 1189 | ||
BC039031.1 | - | 9 | 1305 | 5723005 | ||
FJ200480.1 | - | 8 | 916 | |||
U74649.1 | - | 8 | 1111 | |||
X59770.1 | - | 8 | 1137 |