Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024051.3 | NP_076956.1 | 3 | 504 | ||
NR_037669.1 | 2 | 358 | ||||
GenBank mRNA | AK021779.1 | - | 2 | 983 | ||
AK315608.1 | - | 3 | 495 | |||
BC000625.2 | - | 3 | 499 | 3346822 | ||
BC013937.1 | - | 3 | 465 | 3997629 | ||
BC019243.1 | - | 3 | 496 | 3828006 | ||
BG527593.1 | - | 2 | 347 |