Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024098.3 | NP_077003.1 | 1 | 470 | ||
GenBank mRNA | AK025974.1 | - | 1 | 356 | ||
AK302329.1 | - | 2 | 260 | |||
BC001378.1 | - | 1 | 321 | 3051044 | ||
DA292314.1 | - | 1 | 470 | |||
DQ501251.1 | - | 1 | 301 |