Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322061.1 | NP_001308990.1 | 5 | 968 | ||
NM_001322062.1 | NP_001308991.1 | 4 | 736 | |||
NM_001322064.1 | NP_001308993.1 | 4 | 905 | |||
NM_001322065.1 | NP_001308994.1 | 4 | 780 | |||
NM_001322066.1 | NP_001308995.1 | 4 | 975 | |||
NM_001322067.1 | NP_001308996.1 | 4 | 679 | |||
NM_001322068.1 | NP_001308997.1 | 4 | 804 | |||
NM_001322069.1 | NP_001308998.1 | 4 | 975 | |||
NM_001322070.1 | NP_001308999.1 | 4 | 850 | |||
NM_001322072.1 | NP_001309001.1 | 5 | 1127 | |||
NM_001322073.1 | NP_001309002.1 | 4 | 780 | |||
NM_001322074.1 | NP_001309003.1 | 4 | 907 | |||
NM_001322075.1 | NP_001309004.1 | 4 | 975 | |||
NM_001322076.1 | NP_001309005.1 | 4 | 850 | |||
NM_001322077.1 | NP_001309006.1 | 3 | 394 | |||
NM_024303.2 | NP_077279.1 | 4 | 861 | |||
XM_005259254.1 | XP_005259311.1 | 4 | 736 | |||
XM_006723377.3 | XP_006723440.1 | 3 | 1020 | |||
XM_011527308.1 | XP_011525610.1 | 3 | 545 | |||
XM_017027298.1 | XP_016882787.1 | 3 | 1005 | |||
XM_017027299.1 | XP_016882788.1 | 3 | 1102 | |||
GenBank mRNA | AK098700.1 | - | 3 | 493 | ||
AK223485.1 | - | 2 | 558 | |||
AK301873.1 | - | 3 | 394 | |||
BC002636.1 | - | 3 | 597 | 3606994 | ||
BC043232.1 | - | 5 | 1132 | 5295665 |