Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030763.2 | NP_110390.1 | 3, 4 | 365 | ||
XM_006724688.3 | XP_006724751.1 | 3, 4 | 312 | |||
GenBank mRNA | AF250329.1 | - | 2, 3 | 104 | ||
AK094058.1 | - | 3, 4 | 365 | |||
AK315792.1 | - | 2, 3 | 158 | |||
BC005342.1 | - | 2, 3 | 125 | 3838879 | ||
DA745749.1 | - | 3, 4 | 334 |