Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040451.2 | NP_001035541.1 | 3, 4 | 738 | ||
NM_001040452.2 | NP_001035542.1 | 4, 5 | 549 | |||
NM_025158.4 | NP_079434.3 | 4, 5 | 724 | |||
XM_005265993.4 | XP_005266050.1 | 1, 2 | 95 | |||
XM_006714921.3 | XP_006714984.2 | 4, 5 | 734 | |||
XM_017009891.1 | XP_016865380.1 | 4, 5 | 549 | |||
XM_017009892.1 | XP_016865381.1 | 1, 2 | 113 | |||
XM_017009893.1 | XP_016865382.1 | 1, 2 | 113 | |||
XM_017009894.1 | XP_016865383.1 | 4, 5 | 457 | |||
XM_017009895.1 | XP_016865384.1 | 3, 4 | 529 | |||
GenBank mRNA | AB209507.1 | - | 4, 5 | 449 | ||
AF312367.1 | - | 4, 5 | 715 | |||
AF361055.2 | - | 4, 5 | 697 | |||
AK075021.1 | - | 3, 4 | 637 | |||
AK291926.1 | - | 3, 4 | 738 | |||
BC032571.1 | - | 4, 5 | 468 | 5243223 | ||
DA925727.1 | - | 4, 5 | 510 |