Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CALM2 SIRNA Assays ›
          Gene Symbol:
          CALM2
          Gene Name:
          calmodulin 2
          Gene Aliases:
          CAMII, LQT15, PHKD, PHKD2, caM
          Chromosome Location:
          Chr.2: 47160082 - 47176936 on Build GRCh38
          UniGene ID:
          Hs.468442
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS188460
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CALM2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001305624.1 NP_001292553.1 4 337
          NM_001305625.1 NP_001292554.1 3 196
          NM_001305626.1 NP_001292555.1 2 3551
          NM_001743.5 NP_001734.1 3 291
          GenBank mRNA AK055130.1 - 2 2008
          AK315694.1 - 3 179
          BC003354.1 - 3 167 2899899
          BC006464.1 - 3 169 3542044
          BC008437.1 - 3 185 4092978
          BC017385.1 - 3 183 3681165
          BC018677.1 - 3 184 4656551
          BC026065.1 - 3 183 3683323
          BG563041.1 - 4 231
          BP370358.1 - 3 271
          BT009916.1 - 2 111
          CR541990.1 - 2 111
          CR542021.1 - 2 111
          D45887.1 - 3 179
          HY022313.1 - 3 291
          HY108355.1 - 3 197

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          MAPK cascade
          response to amphetamine
          regulation of heart rate
          platelet degranulation
          detection of calcium ion
          glycogen catabolic process
          muscle contraction
          G-protein coupled receptor signaling pathway
          activation of adenylate cyclase activity
          Wnt signaling pathway, calcium modulating pathway
          positive regulation of peptidyl-threonine phosphorylation
          negative regulation of peptidyl-threonine phosphorylation
          regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          substantia nigra development
          regulation of rhodopsin mediated signaling pathway
          positive regulation of cyclic nucleotide metabolic process
          positive regulation of protein autophosphorylation
          regulation of cytokinesis
          positive regulation of phosphoprotein phosphatase activity
          ion transmembrane transport
          positive regulation of protein dephosphorylation
          Fc-epsilon receptor signaling pathway
          positive regulation of DNA binding
          positive regulation of GTPase activity
          inositol phosphate metabolic process
          regulation of nitric-oxide synthase activity
          positive regulation of nitric-oxide synthase activity
          positive regulation of cyclic-nucleotide phosphodiesterase activity
          response to corticosterone
          response to calcium ion
          regulation of cardiac muscle contraction
          negative regulation of ryanodine-sensitive calcium-release channel activity
          positive regulation of ryanodine-sensitive calcium-release channel activity
          positive regulation of protein serine/threonine kinase activity
          regulation of high voltage-gated calcium channel activity
          regulation of cell communication by electrical coupling involved in cardiac conduction
          Ras guanyl-nucleotide exchange factor activity
          calcium ion binding
          protein binding
          adenylate cyclase binding
          inositol-1,4,5-trisphosphate 3-kinase activity
          ligand-gated ion channel activity
          protein kinase binding
          protein domain specific binding
          nitric-oxide synthase regulator activity
          titin binding
          type 3 metabotropic glutamate receptor binding
          thioesterase binding
          N-terminal myristoylation domain binding
          phospholipase binding
          protein serine/threonine kinase activator activity
          phosphatidylinositol 3-kinase binding
          ion channel binding
          calcium-dependent protein binding
          nitric-oxide synthase binding
          protein phosphatase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0