Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097599.1 | NP_001091068.1 | 2 | 1108 | ||
NM_001097600.1 | NP_001091069.1 | 2 | 959 | |||
NM_025246.2 | NP_079522.2 | 2 | 1373 | |||
XM_006713773.3 | XP_006713836.1 | 2 | 3525 | |||
XM_011513214.2 | XP_011511516.1 | 2 | 1712 | |||
XM_017007289.1 | XP_016862778.1 | 2 | 836 | |||
XM_017007290.1 | XP_016862779.1 | 2 | 1035 | |||
XM_017007291.1 | XP_016862780.1 | 2 | 841 | |||
GenBank mRNA | AK314079.1 | - | 2 | 1268 | ||
AL136661.1 | - | 2 | 1254 | |||
BC000111.1 | - | 2 | 849 | 3508204 | ||
BC022557.1 | - | 2 | 894 | 4793991 | ||
BC050423.1 | - | 2 | 1373 |