Search Thermo Fisher Scientific
Gene Symbol: PRR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077497.2 | NP_001070965.1 | 3 | 1125 | ||
NM_025263.3 | NP_079539.2 | 4 | 1188 | |||
GenBank mRNA | AK074531.1 | - | 3 | 554 | ||
AK131382.1 | - | 2 | 2516 | |||
AK298621.1 | - | 4 | 912 | |||
AK316586.1 | - | 4 | 917 | |||
BC126455.1 | - | 4 | 768 | 8992171 | ||
BC126457.1 | - | 4 | 768 | 8992173 | ||
CN258740.1 | - | 4 | 581 | |||
DR762096.1 | - | 3 | 515 | |||
U63336.1 | - | 4 | 1011 |