Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other THUMPD2 SIRNA Assays ›
Gene Symbol:
THUMPD2
Gene Name:
THUMP domain containing 2
Gene Aliases:
C2orf8
Chromosome Location:
Chr.2: 39736060 - 39779276 on Build GRCh38
UniGene ID:
Hs.468254
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS188479
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: THUMPD2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321468.1
NP_001308397.1
1, 2
161
NM_001321469.1
NP_001308398.1
1, 2
161
NM_001321470.1
NP_001308399.1
1, 2
161
NM_001321474.1
NP_001308403.1
1, 2
161
NM_001321475.1
NP_001308404.1
1, 2
161
NM_001321477.1
NP_001308406.1
1, 2
161
NM_001321478.1
NP_001308407.1
1, 2
161
NM_001321479.1
NP_001308408.1
1, 2
161
NM_001321480.1
NP_001308409.1
1, 2
161
NM_001321481.1
NP_001308410.1
1, 2
161
NM_025264.4
NP_079540.2
1, 2
161
NR_028102.1
1, 2
161
NR_135659.1
1, 2
161
NR_135660.1
1, 2
161
NR_135661.1
1, 2
161
NR_135662.1
1, 2
161
NR_135663.1
1, 2
161
NR_135664.1
1, 2
161
XM_017005050.1
XP_016860539.1
1, 2
161
XM_017005051.1
XP_016860540.1
1, 2
161
XM_017005052.1
XP_016860541.1
1, 2
161
XM_017005053.1
XP_016860542.1
1, 2
161
XM_017005054.1
XP_016860543.1
1, 2
152
GenBank mRNA
AK093580.1
-
1, 2
132
AK226034.1
-
1, 2
111
AK292002.1
-
1, 2
146
AK298178.1
-
1, 2
106
AK301353.1
-
1, 2
37
AK307754.1
-
1, 2
124
AL832115.1
-
1, 2
128
BC004163.2
-
1, 2
122
2961526
BC013299.2
-
1, 2
122
2961526
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA methyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
tRNA methylation
tRNA binding
tRNA (guanine) methyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0