Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMK2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015981.3 | NP_057065.2 | 17 | 1377 | ||
NM_171825.2 | NP_741960.1 | 16 | 1344 | |||
XM_017009898.1 | XP_016865387.1 | 16 | 1344 | |||
GenBank mRNA | AB023185.1 | - | 16 | 1277 | ||
AF091486.1 | - | 16 | 1233 | |||
AF145710.1 | - | 16 | 1135 | |||
AF145711.1 | - | 17 | 1168 | |||
AK289776.1 | - | 17 | 1377 | |||
BC040457.1 | - | 16 | 1259 | 4811948 | ||
BF951952.1 | - | 1 | 34 |