Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203293.2 | NP_976038.1 | 7 | 1254 | ||
NM_203294.1 | NP_976039.1 | 7 | 868 | |||
NM_203295.1 | NP_976040.1 | 7 | 936 | |||
NM_203296.1 | NP_976041.1 | 7 | 1145 | |||
NM_203297.1 | NP_976042.1 | 5 | 1302 | |||
XM_017009903.1 | XP_016865392.1 | 7 | 1293 | |||
GenBank mRNA | AF396651.1 | - | 7 | 1131 | ||
AF396652.1 | - | 7 | 868 | |||
AF396653.1 | - | 7 | 936 | |||
AF396654.1 | - | 7 | 929 | |||
AF396655.1 | - | 5 | 1308 | |||
AK124617.1 | - | 2 | 2655 | |||
BC132863.1 | - | 7 | 1133 | 40146885 | ||
BC132867.1 | - | 7 | 1133 | 40146889 |