Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145646.1 | NP_001139118.1 | 3 | 413 | ||
NM_031301.3 | NP_112591.2 | 3 | 413 | |||
XM_011522105.2 | XP_011520407.1 | 3 | 444 | |||
GenBank mRNA | AB189172.1 | - | 3 | 353 | ||
AB197122.1 | - | 4 | 417 | |||
AF508794.1 | - | 3 | 353 | |||
AK291204.1 | - | 3 | 355 | |||
AK307456.1 | - | 2 | 161 | |||
AK307725.1 | - | 3 | 475 | |||
AK310843.1 | - | 1 | 39 | |||
AL136671.1 | - | 3 | 359 | |||
AY358698.1 | - | 3 | 347 | |||
BC020905.1 | - | 3 | 355 | 4720647 | ||
BI825203.1 | - | 3 | 409 | |||
DA397427.1 | - | 3 | 413 |