Search
Search

MEGF10| Gene Symbol: MEGF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256545.1 | NP_001243474.1 | 10 | 1577 | ||
| NM_001308119.1 | NP_001295048.1 | 11 | 1628 | |||
| NM_001308121.1 | NP_001295050.1 | 10 | 1577 | |||
| NM_032446.2 | NP_115822.1 | 11 | 1628 | |||
| XM_011543694.1 | XP_011541996.1 | 10 | 1340 | |||
| XM_017009987.1 | XP_016865476.1 | 12 | 1873 | |||
| XM_017009988.1 | XP_016865477.1 | 3 | 646 | |||
| GenBank mRNA | AB058676.1 | - | 11 | 1469 | ||
| AK123568.1 | - | 12 | 1743 | |||
| AK307919.1 | - | 10 | 1496 | |||
| BC020198.1 | - | 11 | 1545 | |||
| BC152478.1 | - | 11 | 1469 | 8862616 | ||
| CR749437.1 | - | 10 | 1511 | |||