Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301175.1 | NP_001288104.1 | 11 | 1217 | ||
NM_032663.4 | NP_116052.2 | 8 | 843 | |||
XM_005253962.3 | XP_005254019.1 | 8 | 826 | |||
XM_005253965.4 | XP_005254022.1 | 7 | 654 | |||
XM_006719653.3 | XP_006719716.1 | 9 | 916 | |||
XM_011538894.2 | XP_011537196.1 | 6 | 547 | |||
XM_017020048.1 | XP_016875537.1 | 8 | 838 | |||
XM_017020049.1 | XP_016875538.1 | 8 | 826 | |||
XM_017020050.1 | XP_016875539.1 | 8 | 938 | |||
XM_017020051.1 | XP_016875540.1 | 9 | 794 | |||
XM_017020052.1 | XP_016875541.1 | 8 | 840 | |||
XM_017020053.1 | XP_016875542.1 | 7 | 570 | |||
XM_017020054.1 | XP_016875543.1 | 6 | 445 | |||
GenBank mRNA | AJ586136.1 | - | 8 | 734 | ||
AK027820.1 | - | 8 | 808 | |||
AK097830.1 | - | 11 | 1217 | |||
BC004868.2 | - | 8 | 833 | 3833242 | ||
BC022094.2 | - | 6 | 385 | |||
CN367077.1 | - | 2 | 107 |