Search
Search

SLC35B4| Gene Symbol: SLC35B4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032826.4 | NP_116215.1 | 8 | 977 | ||
| XM_011516645.2 | XP_011514947.1 | 8 | 745 | |||
| GenBank mRNA | AB052892.1 | - | 8 | 953 | ||
| AJ315497.1 | - | 8 | 809 | |||
| AJ315498.1 | - | 8 | 799 | |||
| AJ971941.1 | - | 8 | 653 | |||
| AK027603.1 | - | 8 | 817 | |||
| AK027726.1 | - | 7 | 535 | |||
| AK075368.1 | - | 8 | 782 | |||
| AK222874.1 | - | 8 | 826 | |||
| AK307621.1 | - | 7 | 755 | |||
| BC008413.1 | - | 8 | 798 | 4333393 | ||